missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Diphenylmethylene)aminoacetonitrile 98.0+%, TCI America™
Supplier: TCI America D414525G
Spécifications
| N-(Diphenylmethylene)aminoacetonitrile | |
| 85°C | |
| MFCD00009970 | |
| n-diphenylmethylene aminoacetonitrile, 2-diphenylmethylene amino acetonitrile, benzhydrylidene-amino-acetonitrile, 2-benzhydrylideneamino acetonitrile, 2-diphenylmethylidene amino acetonitrile, acetonitrile, diphenylmethylene amino, 4,4-diphenyl-3-azabut-3-enenitrile, pubchem13883, benzhydrylideneaminoacetonitrile | |
| N#CCN=C(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 220.28 | |
| 220.28 | |
| Crystalline Powder |
| 70591-20-7 | |
| C15H12N2 | |
| 25 g | |
| VRLJFRODHVSTIK-UHFFFAOYSA-N | |
| 2-[(diphenylmethylidene)amino]acetonitrile | |
| 612957 | |
| ≥98.0% (GC) |
Identifiants chimiques
| 70591-20-7 | |
| 220.28 | |
| VRLJFRODHVSTIK-UHFFFAOYSA-N | |
| 612957 | |
| N#CCN=C(C1=CC=CC=C1)C1=CC=CC=C1 |
| C15H12N2 | |
| MFCD00009970 | |
| n-diphenylmethylene aminoacetonitrile, 2-diphenylmethylene amino acetonitrile, benzhydrylidene-amino-acetonitrile, 2-benzhydrylideneamino acetonitrile, 2-diphenylmethylidene amino acetonitrile, acetonitrile, diphenylmethylene amino, 4,4-diphenyl-3-azabut-3-enenitrile, pubchem13883, benzhydrylideneaminoacetonitrile | |
| 2-[(diphenylmethylidene)amino]acetonitrile |
Sécurité et manipulation
missing translation for 'tsca' : No
Recommended Storage : Refrigerator