missing translation for 'onlineSavingsMsg'
Learn More

N-(Diphenylmethylene)aminoacetonitrile 98.0+%, TCI America™

Catalog No. p-7128122
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41455G 5 g
D414525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41455G Supplier TCI America Supplier No. D41455G
Only null left

Chemical Identifiers

CAS 70591-20-7
Molecular Formula C15H12N2
Molecular Weight (g/mol) 220.28
MDL Number MFCD00009970
InChI Key VRLJFRODHVSTIK-UHFFFAOYSA-N
Synonym n-diphenylmethylene aminoacetonitrile, 2-diphenylmethylene amino acetonitrile, benzhydrylidene-amino-acetonitrile, 2-benzhydrylideneamino acetonitrile, 2-diphenylmethylidene amino acetonitrile, acetonitrile, diphenylmethylene amino, 4,4-diphenyl-3-azabut-3-enenitrile, pubchem13883, benzhydrylideneaminoacetonitrile
PubChem CID 612957
IUPAC Name 2-[(diphenylmethylidene)amino]acetonitrile
SMILES N#CCN=C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 85°C
Quantity 5 g
Formula Weight 220.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(Diphenylmethylene)aminoacetonitrile
TSCA No
Recommended Storage Refrigerator