missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 5A, 3-5mm (0.12-0.20in) beads

CAS: 69912-79-4 | AlNaO6Si2 | 202.14 g/mol

Supplier:  Thermo Scientific Chemicals L057220B

 Afficher plus de versions de ce produit

Numéro de catalogue. AAL057220B


Il en reste null
Explore available promotions

Description

Description

Molecular sieves, 13X, 3-5mm (0.12-0.20in) beads are used for liquefied petroleum gas, natural gas drying, desulfurization, clean. It is used as an adsorbent for gases and liquids. It is used in chemistry laboratory for separating compounds and drying reaction starting materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1000 g
Molecular sieves, 5A
69912-79-4
MFCD00147627
URGAHOPLAPQHLN-UHFFFAOYSA-N
aluminum;sodium;dioxido(oxo)silane
19758701
Hygroscopic
3 to 5mm (0.12 to 0.20 in) beads
AlNaO6Si2
Insoluble in water.
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
202.14

Chemical Identifiers

69912-79-4
202.14
URGAHOPLAPQHLN-UHFFFAOYSA-N
aluminum;sodium;dioxido(oxo)silane
AlNaO6Si2
MFCD00147627
19758701
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only