missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 5A, 3-5mm (0.12-0.20in) beads

CAS: 69912-79-4 | AlNaO6Si2 | 202.14 g/mol

$90.37 - $221.10

Chemical Identifiers

CAS 69912-79-4
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.14
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0572230
View Documents
Thermo Scientific Chemicals
L0572230
250 g
Each for $90.37
Only null left
 
AAL057220B
View Documents
Thermo Scientific Chemicals
L057220B
1000 g
Each for $221.10
Only null left
 
Description

Description

Molecular sieves, 13X, 3-5mm (0.12-0.20in) beads are used for liquefied petroleum gas, natural gas drying, desulfurization, clean. It is used as an adsorbent for gases and liquids. It is used in chemistry laboratory for separating compounds and drying reaction starting materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

69912-79-4
202.14
URGAHOPLAPQHLN-UHFFFAOYSA-N
aluminum;sodium;dioxido(oxo)silane
AlNaO6Si2
MFCD00147627
19758701
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

250 g
69912-79-4
MFCD00147627
URGAHOPLAPQHLN-UHFFFAOYSA-N
aluminum;sodium;dioxido(oxo)silane
19758701
Hygroscopic
AlNaO6Si2
Insoluble in water.
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
202.14
Molecular sieves, 5A, 3 to 5mm (0.12 to 0.20 in) beads
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only