Learn More
Invitrogen™ PIP Strips™ Membranes
Identify proteins possessing phosphoinositide domains
$361.00 - $1,152.00
Spécifications
| Content And Storage | Store in refrigerator 2°C to 8°C and protect from light. |
|---|---|
| Product Type | Phospholipid Arrays |
| Format | Membrane |
| Product Line | PIP Strips |
| Shipping Condition | Room Temperature |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
P23750
|
Invitrogen™
P23750 |
2 strips |
chaque for $361.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
P23751
|
Invitrogen™
P23751 |
10 strips |
chaque for $1,152.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
PIP Strips membranes are designed for the identification of proteins possessing phosphoinositide domains and for analysis of their lipid-binding specificities. PIP Strips membranes facilitate the analysis of phosphoinositide protein interactions by protein-lipid overlay assays. Proteins may be detected using standard western blot procedures in conjunction with high-performance alkaline phosphatase- and horseradish peroxidase (HRP)-mediated signal generation systems.
Cell Analysis, Cell Structure, Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Membranes (General) and Lipids, Plasma Membrane, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Spécifications
| Store in refrigerator 2°C to 8°C and protect from light. | |
| Membrane | |
| Room Temperature |
| Phospholipid Arrays | |
| PIP Strips |
For Research Use Only. Not for use in diagnostic procedures.