missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PIP Strips™ Membranes

Catalog No. P23750
Encompass
Change view
Click to view available options
Quantity:
10 strips
2 strips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23750 2 strips
P23751 10 strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23750 Supplier Invitrogen™ Supplier No. P23750

Identify proteins possessing phosphoinositide domains

PIP Strips membranes are designed for the identification of proteins possessing phosphoinositide domains and for analysis of their lipid-binding specificities. PIP Strips membranes facilitate the analysis of phosphoinositide protein interactions by protein-lipid overlay assays. Proteins may be detected using standard western blot procedures in conjunction with high-performance alkaline phosphatase- and horseradish peroxidase (HRP)-mediated signal generation systems.

Cell Analysis, Cell Structure, Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Membranes (General) and Lipids, Plasma Membrane, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Product Type Phospholipid Arrays
Format Membrane
Product Line PIP Strips
Quantity 2 strips
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.