missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ MET-SimDis Capillary GC Column, df 2.65 μm

Designed for simulated distillation, an application performed in the petroleum industry that reveals the boiling point distribution of a sample.

Supplier:  MilliporeSigma™ Supelco™ 29898U

 Afficher plus de versions de ce produit

Numéro de catalogue. 111007589


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Application: This rugged metal column is designed for simulated distillation, an application performed in the petroleum industry that reveals the boiling point distribution of a sample.USP Code: None. Phase: Bonded. Proprietary Temp. Limits:. 2 μm: -25 C to 400 C (isothermal or programmed)
Spécifications

Spécifications

0.53 mm
Capillary GC
Gas Chromatography (GC)
10 m
Bonded
None
-25°C to +400°C (isothermal or programmed)
2.65 μm
Metal
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions