Learn More
Mefloquine hydrochloride, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 461130010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Mefloquine hydrochloride | |
Authentic | |
MFCD00797519,MFCD00797519 | |
mefloquine hcl, r-2,8-bis trifluoromethyl quinolin-4-yl s-piperidin-2-yl methanol hydrochloride, r*,s*-2,8-bis-trifluoromethyl-quinolin-4-yl-piperidin-2-yl-methanol hydrochloride, r-2,8-bis trifluoromethyl quinolin-4-yl-2s-piperidin-2-yl methanol;hydrochloride, as-rel-a-2r-2-piperidinyl-2,8-bis trifluoromethyl-4-quinolinemethanol monohydrochloride, +-mefloquine hydrochloride, unii-5y9l3636o3 component, r-2,8-bis trifluoromethyl-4-quinolinyl 2s-2-piperidinyl methanol hydrochloride, r-2,8-bis trifluoromethyl quinolin-4-yl 2s-piperidin-2-yl methanol hydrochloride, s-2,8-bis trifluoromethyl quinolin-4-yl r-piperidin-2-yl methanolhydrochloride | |
[H+].[Cl-].OC(C1CCCCN1)C1=C2C=CC=C(C2=NC(=C1)C(F)(F)F)C(F)(F)F | |
414.78 | |
414.77 |
51773-92-3 | |
C17H17ClF6N2O | |
1g | |
WESWYMRNZNDGBX-UHFFFAOYNA-N | |
hydrogen [2,8-bis(trifluoromethyl)quinolin-4-yl](piperidin-2-yl)methanol chloride | |
18660697 | |
≥97.5% (HPLC) ≥97.5% (on dry substance) (Argentometry) |
Identifiants chimiques
51773-92-3 | |
414.78 | |
WESWYMRNZNDGBX-UHFFFAOYNA-N | |
18660697 | |
[H+].[Cl-].OC(C1CCCCN1)C1=C2C=CC=C(C2=NC(=C1)C(F)(F)F)C(F)(F)F |
C17H17ClF6N2O | |
MFCD00797519,MFCD00797519 | |
mefloquine hcl, r-2,8-bis trifluoromethyl quinolin-4-yl s-piperidin-2-yl methanol hydrochloride, r*,s*-2,8-bis-trifluoromethyl-quinolin-4-yl-piperidin-2-yl-methanol hydrochloride, r-2,8-bis trifluoromethyl quinolin-4-yl-2s-piperidin-2-yl methanol;hydrochloride, as-rel-a-2r-2-piperidinyl-2,8-bis trifluoromethyl-4-quinolinemethanol monohydrochloride, +-mefloquine hydrochloride, unii-5y9l3636o3 component, r-2,8-bis trifluoromethyl-4-quinolinyl 2s-2-piperidinyl methanol hydrochloride, r-2,8-bis trifluoromethyl quinolin-4-yl 2s-piperidin-2-yl methanol hydrochloride, s-2,8-bis trifluoromethyl quinolin-4-yl r-piperidin-2-yl methanolhydrochloride | |
hydrogen [2,8-bis(trifluoromethyl)quinolin-4-yl](piperidin-2-yl)methanol chloride |
Sécurité et manipulation
GHS H Statement Harmful if swallowed.
GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 257-412-
RUO â Research Use Only