Learn More
MAPA™ TriLites 994 Smooth Finish Tripolymer Gloves, 10 in.
Powder free tripolymer blend of neoprene, nitrile and natural rubber gloves protect from chemical splashes in laboratory environments. Available in bag or dispenser box.
$46.22 - $62.20
Spécifications
Certifications/Compliance ASTM D3578, 21 CFR 170-199
Certifications/Compliance (Europe) Cat 3 CE 0334
Chlorinated Yes
Color Purple
Hand Ambidextrous
voir les spécifications
Avertissement:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Produits 8
Numéro de catalogue Numéro du manufacturier. Size Packaging Type Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Size Packaging Type Prix Quantité  
19-048633A
Afficher les documents
MAPA™
34994016
Small 100/Bag
paquet de 100 chaque for $46.22
caisse de 10 paquet for $462.20
Il en reste null
19-111-126A
Afficher les documents
MAPA™
34994026
Small 100/Dispenser box
paquet de 100 chaque for $57.25
caisse de 10 paquet for $515.20
Il en reste null
19-048633B
Afficher les documents
MAPA™
34994017
Medium 100/Bag
paquet de 100 chaque for $60.96
caisse de 10 paquet for $548.60
Il en reste null
19-111-126B
Afficher les documents
MAPA™
34994027
Medium 100/Dispenser box
paquet de 100 chaque for $57.09
caisse de 10 paquet for $513.80
Il en reste null
19-048633C
Afficher les documents
MAPA™
34994018
Large 100/Bag
paquet de 100 chaque for $62.20
caisse de 10 paquet for $559.80
Il en reste null
19-111-126C
Afficher les documents
MAPA™
34994028
Large 100/Dispenser box
paquet de 100 chaque for $58.94
caisse de 10 paquet for $530.40
Il en reste null
19-048633D
Afficher les documents
MAPA™
34994019
X-Large 100/Bag
paquet de 100 chaque for $61.27
caisse de 10 paquet for $551.40
Il en reste null
19-111-126D
Afficher les documents
MAPA™
34994029
X-Large 100/Dispenser box
paquet de 100 chaque for $57.25
caisse de 10 paquet for $515.20
Il en reste null
Description
Description
Powder free tripolymer blend of neoprene, nitrile and natural rubber gloves protect from chemical splashes in laboratory environments. Available in bag or dispenser box.
Comfort of latex with added strength, tear resistance and chemical-splash protection
  • Smooth, lightly halogenated finish for optimum tactility and grip in wet or dry conditions; prevents tape and labels from sticking to gloves
  • Ideal for laboratories, light assembly and sorting, precision parts handling and light maintenance
  • Total powder particle level ≤0.5 μg/glove
  • Ambidextrous
  • Rolled cuff
  • Thickness: 6 mil
  • Length: 10 in. (25.4 cm)
  • Purple
Spécifications
Spécifications
ASTM D3578, 21 CFR 170-199
Yes
Ambidextrous
Tripolymer
6 mil
Type B
Bacteria, Fungi only
Pebbled
TriLItes 994
25.5 cm
Cat 3 CE 0334
Purple
No
Powder Free
0.15 mm
KPT
Rolled
Unlined
10 in.
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'