Learn More
Lanthanum(III) nitrate hexahydrate, 99.9% (REO)
CAS: 10277-43-7 | H12LaN3O15 | 433.01 g/mol
Supplier: Thermo Scientific Chemicals 01291522
Description
Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Lanthanum(III) nitrate hexahydrate | |
10277-43-7 | |
100 g | |
(REO) | |
MFCD00149751 | |
14,5363 | |
Soluble in water,alcohol and acids. | |
O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
433.01 | |
433.01 (324.92 Anhydrous) |
40°C | |
Crystalline | |
99.9% (REO) | |
H12LaN3O15 | |
UN1477 | |
lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate | |
GJKFIJKSBFYMQK-UHFFFAOYSA-N | |
lanthanum(3+);trinitrate;hexahydrate | |
16211478 | |
99.9% |
Identifiants chimiques
10277-43-7 | |
433.01 | |
GJKFIJKSBFYMQK-UHFFFAOYSA-N | |
16211478 | |
O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
H12LaN3O15 | |
MFCD00149751 | |
lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronateAfficher plus de résultats | |
lanthanum(3+);trinitrate;hexahydrate |
Sécurité et manipulation
GHS H Statement
H272-H315-H319-H335
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'einecsNumber' : 233-238-0
missing translation for 'rtecsNumber' : OE5075000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only