Learn More
Thermo Scientific Chemicals HEPES, 0.5M buffer soln., pH 8.0
CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Supplier: Thermo Scientific Chemicals J63002AK
Description
HEPES has been utilized extensively for research and commercial applications in the cell culture and tissue culture industries. It is useful in the pH range 6.8-8.2. HEPES is used as a zwitterionic, all-purpose Goodâs buffer for use in biological research. HEPES is widely used in cell culture, largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration when compared to bicarbonate buffers, which are also commonly used in cell culture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHEPES has been utilized extensively for research and commercial applications in the cell culture and tissue culture industries. It is useful in the pH range 6.8-8.2. HEPES is used as a zwitterionic, all-purpose âGoodâsâ buffer for use in biological research. HEPES is widely used in cell culture, largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration when compared to bicarbonate buffers, which are also commonly used in cell culture.
Solubility
Fully miscible.
Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Spécifications
| HEPES buffer soln. | |
| 0.5 M | |
| Liquid | |
| C8H18N2O4S | |
| Fully miscible. | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 | |
| 238.30 | |
| CHEBI:42334 |
| 7365-45-9 | |
| 8.0 | |
| 250 mL | |
| MFCD00006158 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid | |
| 23831 |
Identifiants chimiques
| 7365-45-9 | |
| 238.30 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| CHEBI:42334 | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 |
| C8H18N2O4S | |
| MFCD00006158 | |
| 23831 | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid |
Sécurité et manipulation
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only