Learn More
Fisherbrand™ SureGrip™ Pipet Tip Racked or Reload System Tips
Full line of disposable pipet tips available in preloaded carriers for fast, easy loading into empty racks
 Afficher plus de versions de ce produit
Numéro de catalogue. 05-408-194
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Full line of disposable pipet tips available in preloaded carriers for fast, easy loading into empty racks
Easy-to-use: just load the carrier into an empty pipet tip rack (reuse your old racks or purchase separately). Carrier locks onto rack with a quick, press-and-snap action. When empty, recycle or dispose of the carrier and reuse the rack.
  • Compatible with most manufacturers' racks
  • Reduces waste by 80% Certified DNase, RNase, Human DNA and PCR inhibitor free
Pipet Tips in Racks
  • All tips are available racked in sturdy, reusable CellRacks
  • Withstand numerous autoclave cycles
  • Keep tips aligned for pipetter mounting
Several Tip Sizes Available
  • 10μL Pipet tip has graduation marks at 2μL and 10μL
  • 10μL Long Reach tip features extended length (45mm) to reach the bottom of tubes without contaminating pipetter shaft, and graduation marks at 2μL, 5μL and 10μL
  • 200μL Pipet tip features a soft-fit collar for easy mounting and ejection, and graduation marks at 10μL, 50μL and 100μL
  • 1250μL Pipet tip features an extended length (103mm), racked on 9mm centers (multichannel compatible), and graduation marks at 200μL, 500μL and 1000μL
Greener Choice Claims
  • Environmental benefits include:
    • Énergie renouvelable
Spécifications
Spécifications
1250 μL
Natural
10.3cm
Refill
Single-use
Yes
Non-sterile
4.05 in.
DNase, RNase, DNA, and PCR inhibitor Free
Standard Length
Universal Fit
SureGrip
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS