Learn More
Applied Biosystems™ Exonuclease I, high concentration, (20 units/μL)
Exonuclease I can be used to measure the endonucleolytic cleavage of covalently closed circular single-stranded DNA reacted with an endonuclease of interest.
Supplier: Applied Biosystems™ 720735KU
Description
Exonuclease I hydrolyzes single-stranded DNA in the 3'→5' direction, releasing. 5'-mononucleotides and leaving the terminal 5'-dinucleotide intact. Hydrolysis is processive and cannot proceed if the 3' terminus is phosphorylated.Order Info
Each
Spécifications
| 20 U/μL | |
| Storage Buffer, Glycine Buffer | |
| Exonuclease I |
| Exonuclease | |
| 5 units |
For Research Use Only. Not for use in diagnostic procedures.