missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ γ-Interferon, Mouse, Lyophilized, Recombinant Protein, E. coli
$306.29 - $819.14
Spécifications
| Formulation | Lyophilized from 0.5X PBS, pH 7.2. |
|---|---|
| Preparation Method | Reconstitute with sterile water to ≥100μg/mL, containing 0.1% HSA or BSA. Further dilute with aqueous buffers just prior to use. |
| Purification Method | RP-HPLC, FPLC, and SDS-PAGE |
| Source | E. coli |
| Immunogen | γ-Interferon |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
40-732-020UG
|
MilliporeSigma™
40732020UG |
20 μg |
chaque for $306.29
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
40-732-0100UG
|
MilliporeSigma™
407320100UG |
100 μg |
chaque for $819.14
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Recombinant, mouse γ-interferon (γ-IFN) expressed in E. coli as a single, non-glycosylated polypeptide containing 134 amino acids. Purified by chromatographic methods. IFN-γ is a type II cytokine produced by activated T cells and natural killer cells. It plays a significant role immunoregulation and has been implicated in the induction of the expression of class II histocompatibility molecules in cells that do not normally express them, resulting in autoimmune disease.Spécifications
| Lyophilized from 0.5X PBS, pH 7.2. | |
| RP-HPLC, FPLC, and SDS-PAGE | |
| γ-Interferon | |
| Mouse | |
| Lyophilized |
| Reconstitute with sterile water to ≥100μg/mL, containing 0.1% HSA or BSA. Further dilute with aqueous buffers just prior to use. | |
| E. coli | |
| RUO | |
| Recombinant | |
| ≥95% |