missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ γ-Interferon, Mouse, Lyophilized, Recombinant Protein, E. coli
Supplier: MilliporeSigma™ 40732020UG
Description
Recombinant, mouse γ-interferon (γ-IFN) expressed in E. coli as a single, non-glycosylated polypeptide containing 134 amino acids. Purified by chromatographic methods. IFN-γ is a type II cytokine produced by activated T cells and natural killer cells. It plays a significant role immunoregulation and has been implicated in the induction of the expression of class II histocompatibility molecules in cells that do not normally express them, resulting in autoimmune disease.Specifications
| Lyophilized from 0.5X PBS, pH 7.2. | |
| RP-HPLC, FPLC, and SDS-PAGE | |
| E. coli | |
| Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for 2 to7 days at 4°C or for up to 4 months at −20°C. Avoid freeze/thaw cycles of solutions. | |
| Mouse | |
| Lyophilized |
| Reconstitute with sterile water to ≥100μg/mL, containing 0.1% HSA or BSA. Further dilute with aqueous buffers just prior to use. | |
| 20 μg | |
| γ-Interferon | |
| RUO | |
| Recombinant | |
| ≥95% |