missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 10 Coverall (all)
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier: DuPont™ PB120SBUXL002500
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Laydown collar
- Open wrists
- Open ankles
- Manufactured under specifications that do not contain natural rubber latex
Specifications
| General Purpose Coveralls | |
| Unisex | |
| Blue | |
| Elastic | |
| Open | |
| Unhooded | |
| Serged | |
| Laydown | |
| General Maintenance, Janitorial, Cleaning |
| X-Large | |
| ProShield | |
| Single-use | |
| Open | |
| Collared | |
| Front Zipper | |
| Non-sterile | |
| PB120S | |
| 25/Cs. |