missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Desonide

$68.28 - $68.28

Identifiants chimiques

CAS 638-94-8
Molecular Formula C24H32O6
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AC467452500
Afficher les documents
Thermo Scientific Chemicals
467452500
250 mg
chaque for $68.28
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

638-94-8
C24H32O6

Spécifications

638-94-8
275.0°C
416.51
CC1(C)O[C@@H]2C[C@H]3C4CCC5=CC(=O)C=CC5(C)[C@H]4C(O)CC3(C)[C@@]2(O1)C(=O)CO
416.51
250 mg
C24H32O6
WBGKWQHBNHJJPZ-YWZQBGSISA-N
(2S,4R,8S,12S)-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
Desonide
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 211-351-6

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only