Learn More
Sartorius Cubis™ II 36S High-Capacity Essential Micro Balance
The Cubis™ II range of high-capacity micro balances with a maximum load between 32 g and 111 g and a readability between 0.001 mg and 0.002 mg provide the ideal choice for a broad range of applications.
Supplier:  Sartorius MCE36S3S00DHWL
 Afficher plus de versions de ce produit
Numéro de catalogue. 01-675-442
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Découvrez plus d'offres spéciales
Description
Description
  • Ultra-high resolution with up to 61 million weighing steps (digits)
  • End-to-end data integrity. 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management
  • Cleanability, part of compliance
  • Error-free operation. Individual QApp workflows, motorized auto-leveling
  • Allow upgrades for hardware features like automated draft shields or built-in ionizer
  • Optional inner draft shield for best weighing performance
  • Housing: Stainless steel 1.4401, 1.4404, Aluminum, Plastic PBT, PA, Float glass Optiwhite, Control Unit: Aluminum, painted, Plastic PBT, PP, Float glass
Spécifications
Spécifications
USB A, USB B, USB C
Color TFT Touchscreen
Stainless Steel
3.5 sec.
100/240 V
32 g
0.001 mg
HWL Configuration with Draft Shield
Micro Balance
No
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS