missing translation for 'onlineSavingsMsg'
Learn More

Copper wire, 0.5mm (0.02in) dia, annealed, 99.9% (metals basis)

Form: approx. 1.75g/m; Temper: soft | CAS: 7440-50-8 | Cu | 63.55 g/mol

$275.60 - $771.34

Identifiants chimiques

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AA10783H4
Afficher les documents
Thermo Scientific Chemicals
010783H4
100 m
chaque for $275.60
Il en reste null
 
AA10783H7
Afficher les documents
Thermo Scientific Chemicals
010783H7
250 m
chaque for $547.63
Il en reste null
 
AA10783H9
Afficher les documents
Thermo Scientific Chemicals
010783H9
500 m
chaque for $771.34
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications
Afficher plus de résultats
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 231-159-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only