missing translation for 'onlineSavingsMsg'
Learn More

Cesium sulfate, 99.95% (metals basis)

CAS: 10294-54-9 | Cs2O4S | 361.87 g/mol

Supplier:  Thermo Scientific Chemicals 01002714

 Afficher plus de versions de ce produit

Numéro de catalogue. AA1002714


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Cesium sulfate is used to prepare dense aqueous solutions, which finds application in isopycnic centrifugation especially for DNA purification and recovery. It acts as a catalyst for sulfur dioxide oxidation with vanadium or vanadium pentoxide. Further, it acts as a metal oxide catalyst enhancer for heterogeneous processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications
Afficher plus de résultats
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H361f-H373

missing translation for 'einecsNumber' : 233-662-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only