Learn More
Brady™ Long Body Keyed-Different Safety Padlocks with 1.5 in. Shackle Locks
Padlock offers unique keyway which breaks electrical current from passing padlock shackle to cylinder
Supplier:  Brady™ 123406
 Afficher plus de versions de ce produit
Numéro de catalogue. 19-103-489
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Lock with 7.6cm (3 in.) long body provides ample space for identification information
  • Made of nylon plastic
  • Lock labels are supplied with English and Spanish or English and French “DO NOT OPERATE” legends
  • Custom keying and engraving is available
  • Available in a variety of colors and packaging
Greener Choice Claims
  • Environmental benefits include:
    • Énergie renouvelable
Spécifications
Spécifications
Blue
Keyed Different
Nylon
1 Ea.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS