missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories Inc A302149AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
Component of the PeBoW complex, which is required for maturation of 28S and 5.8S ribosomal RNAs and formation of the 60S ribosome.Spécifications
| BOP1 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| P97452, Q14137 | |
| Bop1 | |
| Between 350 and 400 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| BOP1 | |
| block of proliferation 1 protein, ribosome biogenesis protein BOP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12181, 23246 | |
| 4° C | |
| Liquid |