missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$344.88 - $344.88
Specifications
| Antigen | BOP1 |
|---|---|
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
Component of the PeBoW complex, which is required for maturation of 28S and 5.8S ribosomal RNAs and formation of the 60S ribosome.Specifications
| BOP1 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| block of proliferation 1 protein, ribosome biogenesis protein BOP1 | |
| Between 350 and 400 | |
| Primary | |
| 4° C | |
| BOP1 |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P97452, Q14137 | |
| 12181, 23246 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |