missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BIS-TRIS propane, 0.2M buffer soln., pH 6.5

CAS: 64431-96-5 | C11H26N2O6 | 282.337 g/mol

$173.36 - $270.10

Identifiants chimiques

CAS 64431-96-5
Molecular Formula C11H26N2O6
Molecular Weight (g/mol) 282.337
MDL Number MFCD00004689
InChI Key HHKZCCWKTZRCCL-UHFFFAOYSA-N
PubChem CID 125132
ChEBI CHEBI:40947
IUPAC Name 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAJ62354AE
Afficher les documents
Thermo Scientific Chemicals
J62354AE
100 mL
chaque for $173.36
Il en reste null
 
AAJ62354AK
Afficher les documents
Thermo Scientific Chemicals
J62354AK
250 mL
chaque for $270.10
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

64431-96-5
282.337
HHKZCCWKTZRCCL-UHFFFAOYSA-N
CHEBI:40947
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
C11H26N2O6
MFCD00004689
125132
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol

Spécifications

64431-96-5
6.5
100 mL
MFCD00004689
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
282.337
CHEBI:40947
0.2 M
Liquid
C11H26N2O6
HHKZCCWKTZRCCL-UHFFFAOYSA-N
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
125132
BIS-TRIS propane buffer soln.
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only