missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals BIS-TRIS, 0.2M buffer soln., pH 7.4
CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol
Supplier: Thermo Scientific Chemicals J62209AK
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| BIS-TRIS | |
| Colorless | |
| 7.5 | |
| 250 mL | |
| Odorless | |
| OWMVSZAMULFTJU-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol | |
| 81462 |
| 6976-37-0 | |
| 0.2 M | |
| Liquid | |
| C8H19NO5 | |
| MFCD00002853 | |
| C(CO)N(CCO)C(CO)(CO)CO | |
| 209.242 | |
| CHEBI:41250 |
Identifiants chimiques
| 6976-37-0 | |
| 209.242 | |
| OWMVSZAMULFTJU-UHFFFAOYSA-N | |
| CHEBI:41250 | |
| C(CO)N(CCO)C(CO)(CO)CO |
| C8H19NO5 | |
| MFCD00002853 | |
| 81462 | |
| 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
Sécurité et manipulation
TSCA : Yes
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only