missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) ether, 99%, extra pure

CAS: 111-96-6 | C6H14O3 | 134.18 g/mol

Supplier:  Thermo Scientific Chemicals 125850010

 Afficher plus de versions de ce produit

Numéro de catalogue. AC125850010



Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications
Afficher plus de résultats
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
May damage fertility or the unborn child.
Flammable liquid and vapor.
May form explosive peroxides.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Use personal protective equipment as required.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-924-4

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only