Learn More
Labconco™ Biosafety Cabinet Sash Stop
For biosafety cabinets with 8, 10, or 12 inch sashes
Supplier:  Labconco™ 3620302
 Afficher plus de versions de ce produit
Numéro de catalogue. 3620202
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Sash Stop restricts sash from closing to maintain continuous operation.
  • For use with Labconco Class II Biosafety Cabinets.
Spécifications
Spécifications
Purifier Class II Biosafety Cabinets
1 in.
25.4 cm
2.5 cm
10 in.
1 in.
2.5 cm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS