Learn More
Thermo Scientific™ BioBasic™ 18 HPLC Columns
Use this 300AÌ wide-pore silica for the separation of small to medium proteins and other biomolecules.
Supplier: Thermo Scientific™ 72105102130
Description
Separate small to medium peptides with Thermo Scientific™ BioBasic™ 18 LC Columns.
- 300Å pore size for maximum performance with biomolecules
- High peak capacity stationary phase
- Outstanding reproducibility, efficiency and column lifetime
- Excellent for LC/MS separations
Spécifications
| Analytical Column | |
| 100 mm | |
| 5 μm | |
| 300 Å | |
| 60°C | |
| C18 | |
| Spherical, Fully Porous, Ultrapure Silica | |
| Reversed Phase | |
| Reversed Phase | |
| Bio-LC |
| 2.1 mm | |
| 5800 psi (400 bar) | |
| 2 to 9 | |
| 100 m²/g | |
| L1 | |
| BioBasic | |
| 9% | |
| Yes | |
| Analytical Column |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.