missing translation for 'onlineSavingsMsg'
Learn More

BD BD™ Difco™ Dextrose Glucose, Anhydrous

Used with microbiological culture media. BD™ Difco™ Dextrose Glucose, Anhydrous is a form of D(+)-Glucose. The dehydrated form provides greater convenience for the laboratorian and bioprocessing specialist.

$138.87 - $1121.95

Spécifications

Certifications/Compliance ISO/DIS, 11014
Color Whitish
Content And Storage <30°C
Additive Dextrose Glucose
Form Anhydrous Solid
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Type Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Type Prix Quantité  
DF0155-17-4
Afficher les documents
BD
215530
500 g Bottle
chaque for $138.87
Il en reste null
 
DF0155-08-5
Afficher les documents
BD
215520
10 kg Pail
chaque for $1,121.95
Il en reste null
 
Description

Description

D-Glucose is a simple sugar that circulates in the blood of animals as blood sugar, and is made in plants during photosynthesis. It is an important source of power for cellular respiration. Glucose is stored as as a starch in plants and as glycogen in animals.

  • CAS: 50-99-7
  • EC No.: 200-075-1
  • Chemical formula: C6H12O6
  • Molar mass: 180.16g/mol
  • Synonym: Blood Sugar
Spécifications

Spécifications

ISO/DIS, 11014
<30°C
Anhydrous Solid
Whitish
Dextrose Glucose
Microbiological culture media
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'