missing translation for 'onlineSavingsMsg'
Learn More

BD BD™ Difco™ Dextrose Glucose, Anhydrous

Used with microbiological culture media. BD™ Difco™ Dextrose Glucose, Anhydrous is a form of D(+)-Glucose. The dehydrated form provides greater convenience for the laboratorian and bioprocessing specialist.

Supplier:  BD 215530

 View more versions of this product

Catalog No. DF0155-17-4


Only null left
Explore available promotions

Description

Description

D-Glucose is a simple sugar that circulates in the blood of animals as blood sugar, and is made in plants during photosynthesis. It is an important source of power for cellular respiration. Glucose is stored as as a starch in plants and as glycogen in animals.

  • CAS: 50-99-7
  • EC No.: 200-075-1
  • Chemical formula: C6H12O6
  • Molar mass: 180.16g/mol
  • Synonym: Blood Sugar
Specifications

Specifications

ISO/DIS, 11014
<30°C
Anhydrous Solid
Bottle
Whitish
Dextrose Glucose
Microbiological culture media
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions