missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Multi Well High Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
Supplier: Axygen™ RESMW12HP
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Spécifications
| ANSI/SLAS, RNase/DNase-free, non-pyrogenic | |
| 84 mL | |
| Non-sterile | |
| 25/Cs. | |
| 25/Cs. | |
| 12 |
| Clear | |
| Polypropylene | |
| Reagent Reservoir | |
| 25/Pk. | |
| 22 mL | |
| Single-use |