missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Multi Well High Profile Reagent Reservoirs

Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
Specifications
| Certifications/Compliance | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
| Color | Clear |
| Capacity (Metric) | 264 mL |
| Material | Polypropylene |
| Sterility | Sterile |
| Type | Reagent Reservoir |
| No. per Case | 25/Cs. |
| No. per Pack | 25/Pk. |
| Packaging | Individually Wrapped; 25/Cs. |
| Volume (Metric) Well | 22 mL |
| Show More |