Learn More
Axygen™ Maximum Recovery™ Polypropylene Pipette Tip
For ultra-low retention and high yield
$203.43 - $209.87
Spécifications
| Sterility | Sterile |
|---|---|
| Clean Claims | DNase, RNase, and Pyrogen Free |
| Format | Rack |
| For Use With (Equipment) | Universal Fit |
| Product Line | Maxymum Recovery |
| Numéro de catalogue | Numéro du manufacturier. | Volume (Metric) | Color | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Volume (Metric) | Color | Prix | Quantité | ||||
|
14-222-715
|
Axygen™
T200CLRS |
1 to 200 μL | Clear |
paquet de 960 chaque for $203.43
caisse de 5 paquet for $1,017.15
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
14-222-740
|
Axygen™
T300LRS |
0.5 to 10 μL | Clear |
paquet de 960 chaque for $209.87
caisse de 5 paquet for $1,049.35
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
- These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
- Nonpyrogenic
- Beveled
- Maxymum Recovery technology for ultra-low retention and high yield
- Sterile
Spécifications
| Sterile | |
| Rack | |
| Maxymum Recovery |
| DNase, RNase, and Pyrogen Free | |
| Universal Fit | |
| Polypropylene |