missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Maximum Recovery™ Polypropylene Pipette Tip

Catalog No. 14222740
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 to 200 μL
Color:
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
14222740 0.5 to 10 μL Clear
14222715 1 to 200 μL Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14222740 Supplier Axygen™ Supplier No. T300LRS
Only null left

For ultra-low retention and high yield

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
  • Maxymum Recovery technology for ultra-low retention and high yield
  • Sterile

Specifications

Description Non-Filtered
Volume (Metric) 0.5 to 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Clean Claims DNase, RNase, and Pyrogen Free
For Use With (Application) With almost all brands of single and multichannel pipettes
Format Rack
Packaging 96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
For Use With (Equipment) Universal Fit
Product Line Maxymum Recovery
Material Polypropylene
Show More Show Less