Learn More
Ammonium Hydroxide, Certified, 1.00N ±0.02N (1.0M), LabChem™
Supplier: LabChem LC110802
Spécifications
| Ammonium Hydroxide | |
| 1336-21-6,7732-18-5 | |
| H5NO | |
| MFCD00066650 | |
| 0.99g/mL | |
| Soluble in water | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| amine hydrate | |
| 14923 | |
| 35.05 | |
| Passes Test | |
| Poly Bottle | |
| Traceable to NIST |
| Colorless | |
| 98.28,1.72 | |
| NH4OH | |
| 1.00N ±0.02N (1.0M) | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| 1 L | |
| N.O | |
| 35.05 | |
| CHEBI:18219 | |
| Certified | |
| Ammonia | |
| 0.99g/mL | |
| Liquid |
Identifiants chimiques
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| 14923 | |
| amine hydrate |
| H5NO | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| CHEBI:18219 | |
| N.O |
Sécurité et manipulation
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life.
GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash skin with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Warning
Recommended Storage : Room Temperature