missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, 2M solution in methanol, AcroSeal™

CAS: 7664-41-7 | H3N | 17.03 g/mol

$183.37 - $183.37

Spécifications

Chemical Name or Material Ammonia
Color Colorless
CAS 7664-41-7,67-56-1
Molecular Formula H3N
Linear Formula NH3
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC368391000
Afficher les documents
Thermo Scientific Chemicals
368391000
100 mL AcroSeal™ Glass Bottle
chaque for $183.37
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications
Afficher plus de résultats
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only