Learn More
Invitrogen™ Tris (1 M), pH 8.0, RNase-free DFS Item
Invitrogen Molecular biology grade, 1M Tris, pH 8.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use.
Supplier:  Invitrogen™ AM9856
 Afficher plus de versions de ce produit
Numéro de catalogue. AM9856
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Invitrogen Molecular biology grade, 1M Tris, pH 8.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Spécifications
Spécifications
1 M
Liquid
500 mL
Molecular Biology
8
Ambion
1,3-Propanediol, 2-Amino-2-(Hydroxymethyl)-,Hydrochloride
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.