missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Adherent Cell Electrodes

Adherent Cell Electrodes are reusable plate electrodes designed to directly electroporate adherent cells or electroporate cultured cell monolayers on filters.

Supplier:  BTX™ 450531

 Afficher plus de versions de ce produit

Numéro de catalogue. BTX450531


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • Specially designed to electroporate cultured monolayers of cells on laminin-coated filters, or on 12 mm polycarbonate or polyester transwell filter units (0.4 mm pore size)
  • Can directly electroporate adherent cells such as human corneal keratocytes, Caco-2, dental pulp cells, NC65 and BCEC, and MDCK cells
  • Consists of two parallel plate electrodes, 7 x 19 mm with 5 mm gap spacing between the plates
  • Each plate electrode has a 0.3 mm height insulated foot at both ends of the bottom
  • Compatible with BTX™ Systems such as Gemini X2 and ECM™ 2001+
Spécifications

Spécifications

Mammalian Cell Transfections, Plant and Yeast Applications, and Direct Electroporation Onto Adherent Cells Either Growing on Transwell Filters or Coverslips, or Directly Onto Monolayers Without Disruption
Electrode Kit
Adherent Cell Electrode Kit, 5 mm gap, includes 45-0204 cable
Cell Electrode, 450204 Cable
1 Ea.
0.5 to 3.0 mL
Banana Cables
ECM™ 830, ECM™ 2001+, Legacy ECM™ 2001, Gemini X2
300 V
Homogeneous
1 μsec.to 35 msec.
5 mm
Non-sterile
Square Wave
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions