missing translation for 'onlineSavingsMsg'
Learn More

Acetate Standard, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$282.50 - $523.65

Identifiants chimiques

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]
voir les spécifications

Comprend: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
ASACE92Y
Afficher les documents
SPEX CertiPrep Group LLC
ASACE92Y
125 mL Triple-leached LDPE bottle
chaque for $282.50
Il en reste null
 
ASACE92X
Afficher les documents
SPEX CertiPrep Group LLC
ASACE92X
500 mL Triple-leached LDPE bottle
chaque for $523.65
Il en reste null
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
CH3COO-1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Spécifications

Identifiants chimiques

6131-90-4
136.079
23665404
sodium;acetate;trihydrate
C2H9NaO5
AYRVGWHSXIMRAB-UHFFFAOYSA-M
CHEBI:32138
CC(=O)[O-].O.O.O.[Na+]

Spécifications

Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Sodium Acetate,distilled water,conductivity or of similar purity
Ion Chromatography
Standard Solution
AYRVGWHSXIMRAB-UHFFFAOYSA-M
CC(=O)[O-].O.O.O.[Na+]
sodium;acetate;trihydrate
125 mL
CHEBI:32138
Acetate Standard, For Ion Chromatography
6131-90-4,7732-18-5
0.1,99.9
C2H9NaO5
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Certified Reference Material
Sodium Acetate
Water
136.079
23665404
1000mg/L
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'dotInformation' : Nonhazardous

Recommended Storage : Ambient

FDS
missing translation for 'documents'

missing translation for 'documents'

Product intended for laboratory use only