missing translation for 'onlineSavingsMsg'
Learn More

Acetate Standard, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC ASACE92X

 View more versions of this product

Catalog No. ASACE92X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
CH&subscript_3;COO&superscript_minus;1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Acetate Standard
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Sodium Acetate,distilled water,conductivity or of similar purity
Ion Chromatography
Standard Solution
AYRVGWHSXIMRAB-UHFFFAOYSA-M
CC(=O)[O-].O.O.O.[Na+]
sodium;acetate;trihydrate
500 mL
CHEBI:32138
For Ion Chromatography
6131-90-4,7732-18-5
0.1,99.9
C2H9NaO5
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Certified Reference Material
Sodium Acetate
Water
136.079
23665404
1000mg/L

Chemical Identifiers

6131-90-4
136.079
23665404
sodium;acetate;trihydrate
C2H9NaO5
AYRVGWHSXIMRAB-UHFFFAOYSA-M
CHEBI:32138
CC(=O)[O-].O.O.O.[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only