missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide 95.0+%, TCI America™
Supplier: TCI America T3498100MG
Spécifications
| 4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide | |
| 200°C | |
| C15H12N2OS2 | |
| 100 mg | |
| QQGWEXFLMJGCAL-UHFFFAOYSA-N | |
| 4-(4-methylphenyl)sulfanylthieno[2,3-c]pyridine-2-carboxamide | |
| 9839311 | |
| ≥95.0% (HPLC) |
| 251992-66-2 | |
| White-Yellow | |
| MFCD09038566 | |
| 4-p-tolylthio thieno 2,3-c pyridine-2-carboxamide, unii-tp7tb1sszd, tp7tb1sszd, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-carboxamide, 4-p-tolylsulfanyl thieno 2,3-c pyridine-2-carboxamide, thieno 2,3-c pyridine-2-carboxamide, 4-4-methylphenyl thio, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-ca, 4-p-tolylsulfanyl-thieno 2,3-c pyridine-2-carboxylic acid amide | |
| CC1=CC=C(C=C1)SC2=C3C=C(SC3=CN=C2)C(=O)N | |
| 300.394 | |
| 300.39 | |
| Crystalline Powder |
Identifiants chimiques
| 251992-66-2 | |
| 300.394 | |
| QQGWEXFLMJGCAL-UHFFFAOYSA-N | |
| 9839311 | |
| CC1=CC=C(C=C1)SC2=C3C=C(SC3=CN=C2)C(=O)N |
| C15H12N2OS2 | |
| MFCD09038566 | |
| 4-p-tolylthio thieno 2,3-c pyridine-2-carboxamide, unii-tp7tb1sszd, tp7tb1sszd, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-carboxamide, 4-p-tolylsulfanyl thieno 2,3-c pyridine-2-carboxamide, thieno 2,3-c pyridine-2-carboxamide, 4-4-methylphenyl thio, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-ca, 4-p-tolylsulfanyl-thieno 2,3-c pyridine-2-carboxylic acid amide | |
| 4-(4-methylphenyl)sulfanylthieno[2,3-c]pyridine-2-carboxamide |
Sécurité et manipulation
missing translation for 'tsca' : No
Recommended Storage : Refrigerator