missing translation for 'onlineSavingsMsg'
Learn More

4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide 95.0+%, TCI America™

Catalog No. p-7137773
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T349825MG 25 mg
T3498100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T349825MG Supplier TCI America Supplier No. T349825MG
Only null left

Chemical Identifiers

CAS 251992-66-2
Molecular Formula C15H12N2OS2
Molecular Weight (g/mol) 300.394
MDL Number MFCD09038566
InChI Key QQGWEXFLMJGCAL-UHFFFAOYSA-N
Synonym 4-p-tolylthio thieno 2,3-c pyridine-2-carboxamide, unii-tp7tb1sszd, tp7tb1sszd, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-carboxamide, 4-p-tolylsulfanyl thieno 2,3-c pyridine-2-carboxamide, thieno 2,3-c pyridine-2-carboxamide, 4-4-methylphenyl thio, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-ca, 4-p-tolylsulfanyl-thieno 2,3-c pyridine-2-carboxylic acid amide
PubChem CID 9839311
IUPAC Name 4-(4-methylphenyl)sulfanylthieno[2,3-c]pyridine-2-carboxamide
SMILES CC1=CC=C(C=C1)SC2=C3C=C(SC3=CN=C2)C(=O)N

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 25 mg
Formula Weight 300.39
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide
TSCA No
Recommended Storage Refrigerator