Learn More
3-Phenoxybenzyl chloride, 97%
CAS: 53874-66-1 | C13H11ClO | 218.68 g/mol
Supplier: Thermo Scientific Chemicals A1567006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| 3-Phenoxybenzyl chloride | |
| 1.189 | |
| >110°C (230°F) | |
| 1.591 | |
| 5 g | |
| 2212101 | |
| QUYVTGFWFHQVRO-UHFFFAOYSA-N | |
| 1-(chloromethyl)-3-phenoxybenzene | |
| 93291 | |
| 97% |
| 53874-66-1 | |
| 141°C to 142°C (5 mmHg) | |
| C13H11ClO | |
| MFCD00040866 | |
| UN3265 | |
| 3-phenoxybenzyl chloride, 1-chloromethyl-3-phenoxybenzene, m-phenoxybenzyl chloride, benzene, 1-chloromethyl-3-phenoxy, 3-phenoxyphenylmethyl chloride, 3-phenoxybenzylchloride, 3-chloromethyl-1-phenoxybenzene, 1-chlormethyl-3-phenoxybenzol, acmc-1asdb, m-phenoxy-benzyl chloride | |
| C1=CC=C(C=C1)OC2=CC=CC(=C2)CCl | |
| 218.68 | |
| 218.69 |
Identifiants chimiques
| 53874-66-1 | |
| 218.68 | |
| QUYVTGFWFHQVRO-UHFFFAOYSA-N | |
| 93291 | |
| C1=CC=C(C=C1)OC2=CC=CC(=C2)CCl |
| C13H11ClO | |
| MFCD00040866 | |
| 3-phenoxybenzyl chloride, 1-chloromethyl-3-phenoxybenzene, m-phenoxybenzyl chloride, benzene, 1-chloromethyl-3-phenoxy, 3-phenoxyphenylmethyl chloride, 3-phenoxybenzylchloride, 3-chloromethyl-1-phenoxybenzene, 1-chlormethyl-3-phenoxybenzol, acmc-1asdb, m-phenoxy-benzyl chloride | |
| 1-(chloromethyl)-3-phenoxybenzene |
Sécurité et manipulation
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P501c
H302-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 258-831-1
missing translation for 'rtecsNumber' : CZ0790000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only