missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzyl chloride, 97%

Catalog No. p-7022763
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1567003 1 g
AAA1567006 5 g
AAA1567014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1567003 Supplier Thermo Scientific Chemicals Supplier No. A1567003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 53874-66-1 | C13H11ClO | 218.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53874-66-1
Molecular Formula C13H11ClO
Molecular Weight (g/mol) 218.68
MDL Number MFCD00040866
InChI Key QUYVTGFWFHQVRO-UHFFFAOYSA-N
Synonym 3-phenoxybenzyl chloride, 1-chloromethyl-3-phenoxybenzene, m-phenoxybenzyl chloride, benzene, 1-chloromethyl-3-phenoxy, 3-phenoxyphenylmethyl chloride, 3-phenoxybenzylchloride, 3-chloromethyl-1-phenoxybenzene, 1-chlormethyl-3-phenoxybenzol, acmc-1asdb, m-phenoxy-benzyl chloride
PubChem CID 93291
IUPAC Name 1-(chloromethyl)-3-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)CCl

Specifications

Density 1.189
Boiling Point 141°C to 142°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.591
Quantity 1 g
UN Number UN3265
Beilstein 2212101
Formula Weight 218.69
Percent Purity 97%
Chemical Name or Material 3-Phenoxybenzyl chloride
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 258-831-1
RTECSNumber CZ0790000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only