missing translation for 'onlineSavingsMsg'
Learn More

3-n-Hexylthiophene, 98%

CAS: 1693-86-3 | C10H16S | 168.298 g/mol

Supplier:  Thermo Scientific Chemicals H5589114

 Afficher plus de versions de ce produit

Numéro de catalogue. AAH5589114


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.

Solubility
Partly soluble in water.

Notes
Store at room temperature. Avoid strong oxidizing agents.
Spécifications
Afficher plus de résultats
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H301-H311-H226-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only