missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-hydroxybenzoic Acid 97.0+%, TCI America™
Supplier: TCI America A085925G
Spécifications
| 3-Amino-4-hydroxybenzoic Acid | |
| 197°C | |
| MFCD00007697 | |
| benzoic acid, 3-amino-4-hydroxy, cheetan, 3,4-ahba, 3-amino-4-hydroxy-benzoic acid, 5-amino saliciylic acid, 3-amino-4-hydroxy benzoic acid, 3-amino-4-hydroxybenzoic acid hydrochloride, 3-amino-4-hydroxybenzoicacid, 5-amino saliciylic acid 5-asa, pubchem20504 | |
| C1=CC(=C(C=C1C(=O)O)N)O | |
| 153.137 | |
| CHEBI:29476 | |
| ≥97.0% (HPLC,T) |
| 1571-72-8 | |
| C7H7NO3 | |
| 25 g | |
| MRBKRZAPGUCWOS-UHFFFAOYSA-N | |
| 3-amino-4-hydroxybenzoic acid | |
| 65083 | |
| 153.14 | |
| Crystalline Powder |
Identifiants chimiques
| 1571-72-8 | |
| 153.137 | |
| MRBKRZAPGUCWOS-UHFFFAOYSA-N | |
| 65083 | |
| 3-amino-4-hydroxybenzoic acid |
| C7H7NO3 | |
| MFCD00007697 | |
| benzoic acid, 3-amino-4-hydroxy, cheetan, 3,4-ahba, 3-amino-4-hydroxy-benzoic acid, 5-amino saliciylic acid, 3-amino-4-hydroxy benzoic acid, 3-amino-4-hydroxybenzoic acid hydrochloride, 3-amino-4-hydroxybenzoicacid, 5-amino saliciylic acid 5-asa, pubchem20504 | |
| CHEBI:29476 | |
| C1=CC(=C(C=C1C(=O)O)N)O |
Sécurité et manipulation
TSCA : No