missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-hydroxybenzoic Acid 97.0+%, TCI America™

Catalog No. p-7117093
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08595G 5 g
A085925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08595G Supplier TCI America Supplier No. A08595G

Chemical Identifiers

CAS 1571-72-8
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007697
InChI Key MRBKRZAPGUCWOS-UHFFFAOYSA-N
Synonym benzoic acid, 3-amino-4-hydroxy, cheetan, 3,4-ahba, 3-amino-4-hydroxy-benzoic acid, 5-amino saliciylic acid, 3-amino-4-hydroxy benzoic acid, 3-amino-4-hydroxybenzoic acid hydrochloride, 3-amino-4-hydroxybenzoicacid, 5-amino saliciylic acid 5-asa, pubchem20504
PubChem CID 65083
ChEBI CHEBI:29476
IUPAC Name 3-amino-4-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)N)O

Specifications

Melting Point 197°C
Quantity 5 g
Formula Weight 153.14
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-hydroxybenzoic Acid