missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Dodecanone, 95%, Thermo Scientific™
Identifiants chimiques
CAS | 6175-49-1 |
---|---|
Molecular Formula | C12H24O |
Molecular Weight (g/mol) | 184.32 |
MDL Number | MFCD00015064 |
InChI Key | LSKONYYRONEBKA-UHFFFAOYSA-N |
Synonym | 2-dodecanone, methyl decyl ketone, decyl methyl ketone, unii-p5cn8ysv3p, dodecanone-2, p5cn8ysv3p, n-decyl methyl ketone, dodecanone, acmc-1b2ve, 2-dodecanone gcAfficher plus de résultats |
PubChem CID | 22556 |
IUPAC Name | dodecan-2-one |
SMILES | CCCCCCCCCCC(=O)C |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
AC330290250
|
Thermo Scientific Chemicals
330290250 |
25mL | Glass bottle |
N/A
|
N/A | ||||
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
6175-49-1 | |
184.32 | |
LSKONYYRONEBKA-UHFFFAOYSA-N | |
22556 | |
CCCCCCCCCCC(=O)C |
C12H24O | |
MFCD00015064 | |
2-dodecanone, methyl decyl ketone, decyl methyl ketone, unii-p5cn8ysv3p, dodecanone-2, p5cn8ysv3p, n-decyl methyl ketone, dodecanone, acmc-1b2ve, 2-dodecanone gcAfficher plus de résultats | |
dodecan-2-one |
Spécifications
6175-49-1 | |
100.0 | |
247°C | |
Authentic | |
Glass bottle | |
1.4320 to 1.4340 (20°C, 589nm) | |
25mL | |
2-dodecanone, methyl decyl ketone, decyl methyl ketone, unii-p5cn8ysv3p, dodecanone-2, p5cn8ysv3p, n-decyl methyl ketone, dodecanone, acmc-1b2ve, 2-dodecanone gc | |
CCCCCCCCCCC(=O)C | |
184.32 | |
184.32 | |
2-Dodecanone |
94.0 | |
0.8200g/mL | |
108°C | |
94% min. (GC) | |
C12H24O | |
MFCD00015064 | |
0.82 | |
LSKONYYRONEBKA-UHFFFAOYSA-N | |
dodecan-2-one | |
22556 | |
95% |
Sécurité et manipulation
GHS H Statement
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
Collect spillage.
missing translation for 'einecsNumber' : 228-222-5
RUO â Research Use Only