Learn More
2-Bromophenyl isocyanate, 97%
CAS: 1592-00-3 | C7H4BrNO | 198.019 g/mol
Supplier: Thermo Scientific Chemicals L1252106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| 2-Bromophenyl isocyanate | |
| 1.607 | |
| 107°C (224°F) | |
| C7H4BrNO | |
| MFCD00001995 | |
| UN2206 | |
| Moisture sensitive | |
| GOOVAYJIVMBWPP-UHFFFAOYSA-N | |
| 1-bromo-2-isocyanatobenzene | |
| 137097 | |
| 97% |
| 1592-00-3 | |
| 91°C to 92°C (5 mmHg) | |
| Pungent | |
| 1.585 | |
| 5 g | |
| 3588447 | |
| 2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate | |
| C1=CC=C(C(=C1)N=C=O)Br | |
| 198.019 | |
| 198.03 |
Identifiants chimiques
| 1592-00-3 | |
| 198.019 | |
| GOOVAYJIVMBWPP-UHFFFAOYSA-N | |
| 137097 | |
| C1=CC=C(C(=C1)N=C=O)Br |
| C7H4BrNO | |
| MFCD00001995 | |
| 2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate | |
| 1-bromo-2-isocyanatobenzene |
Sécurité et manipulation
GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H331-H334-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only