Learn More
2-Bromophenyl isocyanate, 97%
CAS: 1592-00-3 | C7H4BrNO | 198.019 g/mol
$64.26 - $804.97
Chemical Identifiers
| CAS | 1592-00-3 |
|---|---|
| Molecular Formula | C7H4BrNO |
| Molecular Weight (g/mol) | 198.019 |
| MDL Number | MFCD00001995 |
| InChI Key | GOOVAYJIVMBWPP-UHFFFAOYSA-N |
| Synonym | 2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate |
| PubChem CID | 137097 |
| IUPAC Name | 1-bromo-2-isocyanatobenzene |
| SMILES | C1=CC=C(C(=C1)N=C=O)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1252103
|
Thermo Scientific Chemicals
L1252103 |
1 g |
Each for $64.26
|
|
|||||
|
AAL1252106
|
Thermo Scientific Chemicals
L1252106 |
5 g |
Each for $210.07
|
|
|||||
|
AAL1252114
|
Thermo Scientific Chemicals
L1252114 |
25 g |
Each for $804.97
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1592-00-3 | |
| 198.019 | |
| GOOVAYJIVMBWPP-UHFFFAOYSA-N | |
| 137097 | |
| C1=CC=C(C(=C1)N=C=O)Br |
| C7H4BrNO | |
| MFCD00001995 | |
| 2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate | |
| 1-bromo-2-isocyanatobenzene |
Specifications
| 1592-00-3 | |
| 91°C to 92°C (5 mmHg) | |
| Pungent | |
| 1.585 | |
| 1 g | |
| 3588447 | |
| 2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate | |
| C1=CC=C(C(=C1)N=C=O)Br | |
| 198.019 | |
| 198.03 | |
| 2-Bromophenyl isocyanate |
| 1.607 | |
| 107°C (224°F) | |
| C7H4BrNO | |
| MFCD00001995 | |
| UN2206 | |
| Moisture sensitive | |
| GOOVAYJIVMBWPP-UHFFFAOYSA-N | |
| 1-bromo-2-isocyanatobenzene | |
| 137097 | |
| 97% |
Safety and Handling
GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H331-H334-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only